Anti-PRCP Rat Monoclonal Antibody [clone: 860821]

Supplier: R & D Systems
MAB7164
102886-968EA 577.02 USD
102886-968
Anti-PRCP Rat Monoclonal Antibody [clone: 860821]
Antibodies
Type: Primary
Antigen: PRCP
Clonality: Monoclonal
Clone: 860821
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR