Anti-FGFR2 Rat Monoclonal Antibody [clone: 133730]

Supplier: R & D Systems
MAB7161
220021-471EA 772.25 USD
220021-471
Anti-FGFR2 Rat Monoclonal Antibody [clone: 133730]
Antibodies
Type: Primary
Antigen Symbol: FGFR2
Antigen Name: fibroblast growth factor receptor 2
Antigen Synonyms: AU043015, AW556123, Bek, Fgfr-2, Fgfr-7, Fgfr7, KGFR, KGFRTr, svs
Clonality: Monoclonal
Clone: 133730
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR