Anti-C1RA Rat Monoclonal Antibody [clone: 870332]

Supplier: R & D Systems
MAB7160
102886-966EA 594.37 USD
102886-966
Anti-C1RA Rat Monoclonal Antibody [clone: 870332]
Antibodies
Type: Primary
Antigen: C1RA
Clonality: Monoclonal
Clone: 870332
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR