Anti-RPL17 Mouse Monoclonal Antibody [clone: 702142]

Supplier: R & D Systems
MAB7158
102886-964EA 577.02 USD
102886-964
Anti-RPL17 Mouse Monoclonal Antibody [clone: 702142]
Antibodies
Type: Primary
Antigen: RPL17
Clonality: Monoclonal
Clone: 702142
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR