Anti-DTX1 Mouse Monoclonal Antibody [clone: 730612]

Supplier: R & D Systems
MAB7157
102886-962EA 594.37 USD
102886-962
Anti-DTX1 Mouse Monoclonal Antibody [clone: 730612]
Antibodies
Type: Primary
Antigen: DTX1
Clonality: Monoclonal
Clone: 730612
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR