Anti-TLR7 Rat Monoclonal Antibody [clone: 726606]

Supplier: R & D Systems
MAB7156
102886-960EA 594.37 USD
102886-960
Anti-TLR7 Rat Monoclonal Antibody [clone: 726606]
Antibodies
Type: Primary
Antigen: TLR7
Clonality: Monoclonal
Clone: 726606
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR