Anti-IKBKB Rat Monoclonal Antibody [clone: 725818]

Supplier: R & D Systems
MAB7155
102886-958EA 561.11 USD
102886-958
Anti-IKBKB Rat Monoclonal Antibody [clone: 725818]
Antibodies
Type: Primary
Antigen Symbol: IKBKB
Antigen Name: inhibitor of kappaB kinase beta
Antigen Synonyms: AI132552, IKK-2, IKK-beta, IKK2, IKK[b], IKKbeta
Clonality: Monoclonal
Clone: 725818
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR