Anti-BMP5 Mouse Monoclonal Antibody [clone: 149107]

Supplier: R & D Systems
MAB7151
220021-468EA 772.25 USD
220021-468
Anti-BMP5 Mouse Monoclonal Antibody [clone: 149107]
Antibodies
Type: Primary
Antigen: BMP5
Clonality: Monoclonal
Clone: 149107
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR