Anti-BMP5 Mouse Monoclonal Antibody [clone: 78205]

Supplier: R & D Systems
MAB715
220021-467EA 772.25 USD
220021-467
Anti-BMP5 Mouse Monoclonal Antibody [clone: 78205]
Antibodies
Type: Primary
Antigen: BMP5
Clonality: Monoclonal
Clone: 78205
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR