Anti-NCR3LG1 Mouse Monoclonal Antibody [clone: 875001]

Supplier: R & D Systems
MAB7144
102886-956EA 655.11 USD
102886-956
Anti-NCR3LG1 Mouse Monoclonal Antibody [clone: 875001]
Antibodies
Type: Primary
Antigen: NCR3LG1
Clonality: Monoclonal
Clone: 875001
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR