Anti-GFRA1 Mouse Monoclonal Antibody [clone: 260714]

Supplier: R & D Systems
MAB7141
220021-466EA 594.37 USD
220021-466
Anti-GFRA1 Mouse Monoclonal Antibody [clone: 260714]
Antibodies
Type: Primary
Antigen Symbol: GFRA1
Antigen Name: GDNF family receptor alpha 1
Antigen Synonyms: GDNFR, GDNFRA, GFR-ALPHA-1, RET1L, RETL1, TRNR1
Clonality: Monoclonal
Clone: 260714
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR