Anti-BoNT-E Heavy Chain Mouse Monoclonal Antibody [clone: 785208]

Supplier: R & D Systems
MAB7135
102886-954EA 594.37 USD
102886-954
Anti-BoNT-E Heavy Chain Mouse Monoclonal Antibody [clone: 785208]
Antibodies
Type: Primary
Antigen Symbol:
Antigen Name: BoNT-E Heavy Chain
Antigen Synonyms:
Clonality: Monoclonal
Clone: 785208
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: C. botulinum
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR