Anti-HSF1 Rat Monoclonal Antibody [clone: 744509]

Supplier: R & D Systems
MAB7130
102886-952EA 561.11 USD
102886-952
Anti-HSF1 Rat Monoclonal Antibody [clone: 744509]
Antibodies
Type: Primary
Antigen: HSF1
Clonality: Monoclonal
Clone: 744509
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR