Anti-SHC1 Mouse Monoclonal Antibody [clone: 725001]

Supplier: R & D Systems
MAB7129
102886-950EA 577.02 USD
102886-950
Anti-SHC1 Mouse Monoclonal Antibody [clone: 725001]
Antibodies
Type: Primary
Antigen: SHC1
Clonality: Monoclonal
Clone: 725001
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR