Anti-MAP3K2 Mouse Monoclonal Antibody [clone: 722728]

Supplier: R & D Systems
MAB7128
102886-948EA 577.02 USD
102886-948
Anti-MAP3K2 Mouse Monoclonal Antibody [clone: 722728]
Antibodies
Type: Primary
Antigen: MAP3K2
Clonality: Monoclonal
Clone: 722728
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR