Anti-GAB3 Mouse Monoclonal Antibody [clone: 720306]

Supplier: R & D Systems
MAB7127
102886-946EA 577.02 USD
102886-946
Anti-GAB3 Mouse Monoclonal Antibody [clone: 720306]
Antibodies
Type: Primary
Antigen Symbol: GAB3
Antigen Name: GRB2-associated binding protein 3
Antigen Synonyms: -
Clonality: Monoclonal
Clone: 720306
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR