Anti-SPI1 Rat Monoclonal Antibody [clone: 823123]

Supplier: R & D Systems
MAB7124
102886-940EA 600.15 USD
102886-940
Anti-SPI1 Rat Monoclonal Antibody [clone: 823123]
Antibodies
Type: Primary
Antigen: SPI1
Clonality: Monoclonal
Clone: 823123
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR