Anti-SREBF2 Mouse Monoclonal Antibody [clone: 751512]

Supplier: R & D Systems
MAB7119
102886-938EA 616.06 USD
102886-938
Anti-SREBF2 Mouse Monoclonal Antibody [clone: 751512]
Antibodies
Type: Primary
Antigen: SREBF2
Clonality: Monoclonal
Clone: 751512
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR