Anti-ROBO1 Mouse Monoclonal Antibody [clone: 770502]

Supplier: R & D Systems
MAB71181
102886-936EA 600.15 USD
102886-936
Anti-ROBO1 Mouse Monoclonal Antibody [clone: 770502]
Antibodies
Type: Primary
Antigen: ROBO1
Clonality: Monoclonal
Clone: 770502
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR