Anti-ROBO1 Mouse Monoclonal Antibody [clone: 770506]

Supplier: R & D Systems
MAB7118
102886-206EA 594.37 USD
102886-206
Anti-ROBO1 Mouse Monoclonal Antibody [clone: 770506]
Antibodies
Type: Primary
Antigen: ROBO1
Clonality: Monoclonal
Clone: 770506
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR