Anti-CSF2 Mouse Monoclonal Antibody [clone: 111802]

Supplier: R & D Systems
MAB711
220021-465EA 763.57 USD
220021-465
Anti-CSF2 Mouse Monoclonal Antibody [clone: 111802]
Antibodies
Type: Primary
Antigen: CSF2
Clonality: Monoclonal
Clone: 111802
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR