Anti-ETV5 Mouse Monoclonal Antibody [clone: 654404]

Supplier: R & D Systems
MAB7107
102886-202EA 594.37 USD
102886-202
Anti-ETV5 Mouse Monoclonal Antibody [clone: 654404]
Antibodies
Type: Primary
Antigen: ETV5
Clonality: Monoclonal
Clone: 654404
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR