Anti-LIPE Mouse Monoclonal Antibody [clone: 729923]

Supplier: R & D Systems
MAB7104
102886-934EA 577.02 USD
102886-934
Anti-LIPE Mouse Monoclonal Antibody [clone: 729923]
Antibodies
Type: Primary
Antigen: LIPE
Clonality: Monoclonal
Clone: 729923
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR