Anti-KATNA1 Mouse Monoclonal Antibody [clone: 717705]

Supplier: R & D Systems
MAB7100
102886-932EA 577.02 USD
102886-932
Anti-KATNA1 Mouse Monoclonal Antibody [clone: 717705]
Antibodies
Type: Primary
Antigen: KATNA1
Clonality: Monoclonal
Clone: 717705
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR