Anti-FGFR3 Rat Monoclonal Antibody [clone: 136812]

Supplier: R & D Systems
MAB710
220021-464EA 763.57 USD
220021-464
Anti-FGFR3 Rat Monoclonal Antibody [clone: 136812]
Antibodies
Type: Primary
Antigen Symbol: FGFR3
Antigen Name: fibroblast growth factor receptor 3
Antigen Synonyms: CD333, FR3, Fgfr-3, Flg-2, HBGFR, Mfr3, sam3
Clonality: Monoclonal
Clone: 136812
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR