Anti-KLRE1 Rat Monoclonal Antibody [clone: 854929]

Supplier: R & D Systems
MAB7099
102886-930EA 577.02 USD
102886-930
Anti-KLRE1 Rat Monoclonal Antibody [clone: 854929]
Antibodies
Type: Primary
Antigen: KLRE1
Clonality: Monoclonal
Clone: 854929
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR