Anti-PANX1 Mouse Monoclonal Antibody [clone: 720505]

Supplier: R & D Systems
MAB7097
102886-926EA 633.42 USD
102886-926
Anti-PANX1 Mouse Monoclonal Antibody [clone: 720505]
Antibodies
Type: Primary
Antigen: PANX1
Clonality: Monoclonal
Clone: 720505
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR