Anti-NOD1 Mouse Monoclonal Antibody [clone: 626919]

Supplier: R & D Systems
MAB7090
102886-922EA 566.89 USD
102886-922
Anti-NOD1 Mouse Monoclonal Antibody [clone: 626919]
Antibodies
Type: Primary
Antigen: NOD1
Clonality: Monoclonal
Clone: 626919
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR