Anti-S1PR1 Rat Monoclonal Antibody [clone: 713412]

Supplier: R & D Systems
MAB7089
102886-920EA 655.11 USD
102886-920
Anti-S1PR1 Rat Monoclonal Antibody [clone: 713412]
Antibodies
Type: Primary
Antigen: S1PR1
Clonality: Monoclonal
Clone: 713412
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR