Anti-PANX2 Mouse Monoclonal Antibody [clone: 719406]

Supplier: R & D Systems
MAB7088
102886-918EA 561.11 USD
102886-918
Anti-PANX2 Mouse Monoclonal Antibody [clone: 719406]
Antibodies
Type: Primary
Antigen: PANX2
Clonality: Monoclonal
Clone: 719406
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR