Anti-SULF2 Mouse Monoclonal Antibody [clone: 2B4]

Supplier: R & D Systems
MAB7087
102886-916EA 594.37 USD
102886-916
Anti-SULF2 Mouse Monoclonal Antibody [clone: 2B4]
Antibodies
Type: Primary
Antigen: SULF2
Clonality: Monoclonal
Clone: 2B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR