Anti-GPRASP2 Mouse Monoclonal Antibody [clone: 698742]

Supplier: R & D Systems
MAB7078
102886-912EA 561.11 USD
102886-912
Anti-GPRASP2 Mouse Monoclonal Antibody [clone: 698742]
Antibodies
Type: Primary
Antigen Symbol: GPRASP2
Antigen Name: G protein-coupled receptor associated sorting protein 2
Antigen Synonyms: GASP2
Clonality: Monoclonal
Clone: 698742
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR