Anti-CD1A Mouse Monoclonal Antibody [clone: 703217]

Supplier: R & D Systems
MAB7076
102886-910EA 594.37 USD
102886-910
Anti-CD1A Mouse Monoclonal Antibody [clone: 703217]
Antibodies
Type: Primary
Antigen: CD1A
Clonality: Monoclonal
Clone: 703217
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR