Anti-APPL1 Mouse Monoclonal Antibody [clone: 709208]

Supplier: R & D Systems
MAB7075
102886-908EA 561.11 USD
102886-908
Anti-APPL1 Mouse Monoclonal Antibody [clone: 709208]
Antibodies
Type: Primary
Antigen: APPL1
Clonality: Monoclonal
Clone: 709208
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR