Anti-OCLN Mouse Monoclonal Antibody [clone: 690213]

Supplier: R & D Systems
MAB7074
102886-906EA 594.37 USD
102886-906
Anti-OCLN Mouse Monoclonal Antibody [clone: 690213]
Antibodies
Type: Primary
Antigen: OCLN
Clonality: Monoclonal
Clone: 690213
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR