Anti-CASP3 Mouse Monoclonal Antibody [clone: 84803]

Supplier: R & D Systems
MAB707
220021-463EA 594.37 USD
220021-463
Anti-CASP3 Mouse Monoclonal Antibody [clone: 84803]
Antibodies
Type: Primary
Antigen: CASP3
Clonality: Monoclonal
Clone: 84803
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR