Anti-CSF2RA Mouse Monoclonal Antibody [clone: 31916]

Supplier: R & D Systems
MAB706
220021-462EA 772.25 USD
220021-462
Anti-CSF2RA Mouse Monoclonal Antibody [clone: 31916]
Antibodies
Type: Primary
Antigen: CSF2RA
Clonality: Monoclonal
Clone: 31916
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR