Anti-IDH1 Mouse Monoclonal Antibody [clone: 843219]

Supplier: R & D Systems
MAB7049
102886-904EA 600.15 USD
102886-904
Anti-IDH1 Mouse Monoclonal Antibody [clone: 843219]
Antibodies
Type: Primary
Antigen: IDH1
Clonality: Monoclonal
Clone: 843219
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR