Anti-FADD Mouse Monoclonal Antibody [clone: 709736]

Supplier: R & D Systems
MAB7047
102886-902EA 577.02 USD
102886-902
Anti-FADD Mouse Monoclonal Antibody [clone: 709736]
Antibodies
Type: Primary
Antigen Symbol: FADD
Antigen Name: Fas (TNFRSF6)-associated via death domain
Antigen Synonyms: GIG3, MORT1
Clonality: Monoclonal
Clone: 709736
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR