Anti-CCNA1 Mouse Monoclonal Antibody [clone: 722407]

Supplier: R & D Systems
MAB7046
102886-900EA 594.37 USD
102886-900
Anti-CCNA1 Mouse Monoclonal Antibody [clone: 722407]
Antibodies
Type: Primary
Antigen: CCNA1
Clonality: Monoclonal
Clone: 722407
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR