Anti-CD44 (pan specific) Mouse Monoclonal Antibody [clone: 691534]

Supplier: R & D Systems
MAB7045
102886-898EA 639.2 USD
102886-898
Anti-CD44 (pan specific) Mouse Monoclonal Antibody [clone: 691534]
Antibodies
Type: Primary
Antigen: CD44 (pan specific)
Clonality: Monoclonal
Clone: 691534
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR