Anti-CBLN2 Mouse Monoclonal Antibody [clone: 704323]

Supplier: R & D Systems
MAB70441
102886-896EA 577.02 USD
102886-896
Anti-CBLN2 Mouse Monoclonal Antibody [clone: 704323]
Antibodies
Type: Primary
Antigen: CBLN2
Clonality: Monoclonal
Clone: 704323
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR