Anti-CBLN2 Mouse Monoclonal Antibody [clone: 704330]

Supplier: R & D Systems
MAB7044
102886-894EA 577.02 USD
102886-894
Anti-CBLN2 Mouse Monoclonal Antibody [clone: 704330]
Antibodies
Type: Primary
Antigen: CBLN2
Clonality: Monoclonal
Clone: 704330
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR