Anti-CASP8 Mouse Monoclonal Antibody [clone: 84131]

Supplier: R & D Systems
MAB704
220021-460EA 594.37 USD
220021-460
Anti-CASP8 Mouse Monoclonal Antibody [clone: 84131]
Antibodies
Type: Primary
Antigen: CASP8
Clonality: Monoclonal
Clone: 84131
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR