Anti-FUCA1 Mouse Monoclonal Antibody [clone: 810607]

Supplier: R & D Systems
MAB7039
102886-892EA 577.02 USD
102886-892
Anti-FUCA1 Mouse Monoclonal Antibody [clone: 810607]
Antibodies
Simple Western

Type: Primary
Antigen Symbol: FUCA1
Antigen Name: fucosidase, alpha-L- 1, tissue
Antigen Synonyms: FUCA
Clonality: Monoclonal
Clone: 810607
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR