Anti-SARM1 Mouse Monoclonal Antibody [clone: 754711]

Supplier: R & D Systems
MAB7037
102886-890EA 594.37 USD
102886-890
Anti-SARM1 Mouse Monoclonal Antibody [clone: 754711]
Antibodies
Type: Primary
Antigen: SARM1
Clonality: Monoclonal
Clone: 754711
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR