Anti-SHANK2 Mouse Monoclonal Antibody [clone: 711924]

Supplier: R & D Systems
MAB7035
102886-888EA 577.02 USD
102886-888
Anti-SHANK2 Mouse Monoclonal Antibody [clone: 711924]
Antibodies
Type: Primary
Antigen: SHANK2
Clonality: Monoclonal
Clone: 711924
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR