Anti-UBB Mouse Monoclonal Antibody [clone: 83456]

Supplier: R & D Systems
MAB7031
220021-459EA 772.25 USD
220021-459
Anti-UBB Mouse Monoclonal Antibody [clone: 83456]
Antibodies
Type: Primary
Antigen: UBB
Clonality: Monoclonal
Clone: 83456
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR