Anti-KLRG1 Mouse Monoclonal Antibody [clone: 697213]

Supplier: R & D Systems
MAB70291
102886-886EA 594.37 USD
102886-886
Anti-KLRG1 Mouse Monoclonal Antibody [clone: 697213]
Antibodies
Type: Primary
Antigen: KLRG1
Clonality: Monoclonal
Clone: 697213
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR