Anti-GDPD2 Mouse Monoclonal Antibody [clone: 501016]

Supplier: R & D Systems
MAB7026
102886-884EA 577.02 USD
102886-884
Anti-GDPD2 Mouse Monoclonal Antibody [clone: 501016]
Antibodies
Type: Primary
Antigen Symbol: GDPD2
Antigen Name: glycerophosphodiester phosphodiesterase domain containing 2
Antigen Synonyms: GDE3, OBDPF
Clonality: Monoclonal
Clone: 501016
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR